Learn More
Biomatik Corporation Rat Gelsolin (GSN) ELISA Kit
Supplier:  Biomatik Corporation EKU10863
Encompass_Preferred
Rat Gelsolin (GSN) ELISA Kit for detecting GSN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.135ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q68FP1. Gene ID: 296654. Target Synonyms: GS; AGEL; ADF; Brevin; Gelsolin Amyloidosis,Finnish Type; Actin-depolymerizing factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3232