Learn More
Biomatik Corporation Dog Hepcidin (Hepc) ELISA Kit
Supplier:  Biomatik Corporation EKU10900
Encompass_Preferred
Dog Hepcidin (Hepc) ELISA Kit for detecting Hepc in serum, plasma, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.625-40ng/ml. Sensitivity: 0.244ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5U9D2. Gene ID: 492281. Target Synonyms: HAMP; HFE2B; PLTR; LEAP1; Hepcidin Antimicrobial Peptide; Liver-expressed antimicrobial peptide 1; Putative liver tumor regressor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3236