Learn More
Biomatik Corporation Human Oxytocin Receptor (OXTR) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10956
Encompass_Preferred
Human Oxytocin Receptor (OXTR) High-sensitive ELISA Kit for detecting OXTR in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 30pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P30559. Gene ID: 5021. Target Synonyms: OT-R. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3242