Learn More
Biomatik Corporation Human Microfibrillar Associated Protein 5 (MFAP5) ELISA Kit
Supplier:  Biomatik Corporation EKU11070
Encompass_Preferred
Human Microfibrillar Associated Protein 5 (MFAP5) ELISA Kit for detecting MFAP5 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13361. Gene ID: 8076. Target Synonyms: MAGP2; MP25; Microfibril-associated glycoprotein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3257