Learn More
Biomatik Corporation Human Angiopoietin Like Protein 8 (ANGPTL8) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11077
Encompass_Preferred
Human Angiopoietin Like Protein 8 (ANGPTL8) Mini sample ELISA Kit for detecting ANGPTL8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6UXH0. Gene ID: 55908. Target Synonyms: RIFL; TD26; C19orf80; Lipasin; Betatrophin; Hepatocellular Carcinoma-Associated Protein TD26. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3259