Learn More
Biomatik Corporation Mouse Thymus Expressed Chemokine (TECK) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11257
Encompass_Preferred
Mouse Thymus Expressed Chemokine (TECK) Mini sample ELISA Kit for detecting TECK in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 11.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35903. Gene ID: 20300. Target Synonyms: SCYA25; CCL25; Ckb15; Small Inducible Cytokine Subfamily A (Cys-Cys) Member 25; Ck Beta-15; Thymus Expressed Chemokine; TECKvar. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3273