Learn More
Biomatik Corporation Resveratrol (RVT) ELISA Kit
Supplier:  Biomatik Corporation EKU11399
Encompass_Preferred
Resveratrol (RVT) ELISA Kit for detecting RVT in plant tissue, orther biological fluids. Species Reactivity: Universal. Detection Range: 246.9-20,000ng/ml. Sensitivity: 89.3ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: trans-3,5,4'-Trihydroxystilbene; 3,4',5-Stilbenetriol; trans-Resveratrol. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3286