Learn More
Biomatik Corporation Mouse Urocortin 2 (UCN2) ELISA Kit
Supplier:  Biomatik Corporation EKU11510
Encompass_Preferred
Mouse Urocortin 2 (UCN2) ELISA Kit for detecting UCN2 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.23-100ng/ml. Sensitivity: 0.44ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q99ML8. Gene ID: 171530. Target Synonyms: URP; SRP; UCN-II; UCNII; Stresscopin-Related Peptide; Urocortin-related peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3301