Learn More
Biomatik Corporation Rat WNT1 Inducible Signaling Pathway Protein 1 (WISP1) ELISA Kit
Supplier:  Biomatik Corporation EKU11542
Encompass_Preferred
Rat WNT1 Inducible Signaling Pathway Protein 1 (WISP1) ELISA Kit for detecting WISP1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 11.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99PP0. Gene ID: 65154. Target Synonyms: CCN4; WISP1c; WISP1i; WISP1tc; CCN family member 4; Wnt-1-induced secreted protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3307