Learn More
Biomatik Corporation Chicken Insulin (INS) ELISA Kit
Supplier:  Biomatik Corporation EKU11569
Encompass_Preferred
Chicken Insulin (INS) ELISA Kit for detecting INS in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 123.5-10,000pg/ml. Sensitivity: 50.7pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P67970. Gene ID: 396145. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3311