Learn More
Biomatik Corporation Bacitracin (Bac) ELISA Kit
Supplier:  Biomatik Corporation EKU11587
Encompass_Preferred
Bacitracin (Bac) ELISA Kit for detecting Bac in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 8-5,000pg/ml. Sensitivity: 4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3315