Learn More
Biomatik Corporation Human Interleukin 3 (IL3) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11607
Encompass_Preferred
Human Interleukin 3 (IL3) Mini sample ELISA Kit for detecting IL3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08700. Gene ID: 3562. Target Synonyms: MCGF; MULTI-CSF; MCSF; M-CSF; Multiple; Multilineage-Colony-Stimulating Factor; Hematopoietic Growth Factor; P-Cell Stimulating Factor; Mast-Cell Growth Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3318