Learn More
Biomatik Corporation Mouse Procollagen I N-Terminal Propeptide (PINP) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11631
Encompass_Preferred
Mouse Procollagen I N-Terminal Propeptide (PINP) Mini sample ELISA Kit for detecting PINP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.70-300ng/ml. Sensitivity: 1.44ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11087. Gene ID: 12842. Target Synonyms: P1NP; N-Propeptide Of Type I Procollagen; Procollagen I Amino Terminal Propeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3325