Learn More
Biomatik Corporation Human Glycogen Debranching Enzyme (AGL) ELISA Kit
Supplier:  Biomatik Corporation EKU11736
Encompass_Preferred
Human Glycogen Debranching Enzyme (AGL) ELISA Kit for detecting AGL in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35573. Gene ID: 178. Target Synonyms: GDE; Amylo-1,6-Glucosidase; 4-Alpha-Glucanotransferase; Glycogen debrancher; 4-alpha-glucanotransferase; Oligo-1,4-1,4-glucantransferase; Dextrin 6-alpha-D-glucosidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3336