Learn More
Biomatik Corporation Human Hypoxia Inducible Factor 3 Alpha (HIF3a) ELISA Kit
Supplier:  Biomatik Corporation EKU11742
Encompass_Preferred
Human Hypoxia Inducible Factor 3 Alpha (HIF3a) ELISA Kit for detecting HIF3a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y2N7. Gene ID: 64344. Target Synonyms: HIF-3A; HIF-3A4; IPAS; MOP7; PASD7; Basic-helix-loop-helix-PAS protein MOP7; Class E basic helix-loop-helix protein 17; Inhibitory PAS domain protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3338