Learn More
Biomatik Corporation Human Lysine Specific Demethylase 6B (KDM6B) ELISA Kit
Supplier:  Biomatik Corporation EKU11785
Encompass_Preferred
Human Lysine Specific Demethylase 6B (KDM6B) ELISA Kit for detecting KDM6B in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O15054. Target Synonyms: JMJD3; Jumonji Domain Containing 3; Histone Jysine Demethylase; Lysine demethylase 6B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3345