Learn More
Biomatik Corporation Human Tyrosine 3/Tryptophan 5 Monooxygenase Activation Protein Epsilon (YWHAe) ELISA Kit
Supplier:  Biomatik Corporation EKU11812
Encompass_Preferred
Human Tyrosine 3/Tryptophan 5 Monooxygenase Activation Protein Epsilon (YWHAe) ELISA Kit for detecting YWHAe in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P62258. Gene ID: 7531. Target Synonyms: MDCR; MDS; KCIP-1; 14-3-3E; 14-3-3 protein epsilon. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3349