Learn More
Biomatik Corporation Human PR Domain Containing Protein 16 (PRDM16) ELISA Kit
Supplier:  Biomatik Corporation EKU11837
Encompass_Preferred
Human PR Domain Containing Protein 16 (PRDM16) ELISA Kit for detecting PRDM16 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9HAZ2. Gene ID: 63976. Target Synonyms: MEL1; PFM13; MDS1/EVI1-like; PR-Domain Zinc Finger Protein 16; Transcription Factor MEL1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3354