Learn More
Biomatik Corporation Human Collagen Type III (COL3) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU11876
Encompass_Preferred
Human Collagen Type III (COL3) Anti-antigen ELISA Kit for detecting COL3 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.12ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P02461. Gene ID: 1281. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3360