Learn More
Biomatik Corporation Human Fibroblast Growth Factor 2, Basic (FGF2) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11903
Encompass_Preferred
Human Fibroblast Growth Factor 2, Basic (FGF2) Mini sample ELISA Kit for detecting FGF2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09038. Gene ID: 2247. Target Synonyms: B-FGF; BFGF; FGFB; HBGH-2; Basic Fibroblast Growth Factor; Heparin-binding growth factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3368