Learn More
Biomatik Corporation Human Thioredoxin-interacting Protein (TXNIP) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11949
Encompass_Preferred
Human Thioredoxin-interacting Protein (TXNIP) Mini sample ELISA Kit for detecting TXNIP in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H3M7. Gene ID: 10628. Target Synonyms: TBP2; HHCPA78; THIF; VDUP1;Thioredoxin Binding Protein 2 ; Vitamin D3-Upregulated Protein 1; Upregulated By 1,25-Dihydroxyvitamin D-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3381