Learn More
Biomatik Corporation Simian Cytokeratin Fragment Antigen 21-1 (CYFRA21-1) ELISA Kit
Supplier:  Biomatik Corporation EKU11951
Encompass_Preferred
Simian Cytokeratin Fragment Antigen 21-1 (CYFRA21-1) ELISA Kit for detecting CYFRA21-1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Simian. Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A0A2K5W9Q7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3383