Learn More
Biomatik Corporation Human Neurofibromin 1 (NF1) ELISA Kit
Supplier:  Biomatik Corporation EKU12005
Encompass_Preferred
Human Neurofibromin 1 (NF1) ELISA Kit for detecting NF1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 30pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P21359. Gene ID: 4763. Target Synonyms: NFNS; VRNF; WSS; Neurofibromatosis; Von Recklinghausen disease; Watson disease; Neurofibromatosis-related protein NF-1; Neurofibromin truncated. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3398