Learn More
Biomatik Corporation Human Low Density Lipoprotein Receptor Related Protein 2 (LRP2) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12015
Encompass_Preferred
Human Low Density Lipoprotein Receptor Related Protein 2 (LRP2) Mini sample ELISA Kit for detecting LRP2 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P98164. Gene ID: 4036. Target Synonyms: DBS; gp330; Glycoprotein 330; Megalin; Low Density Lipoprotein-Related Protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3401