Learn More
Biomatik Corporation Mouse Homogentisate-1,2-Dioxygenase (HGD) ELISA Kit
Supplier:  Biomatik Corporation EKU12033
Encompass_Preferred
Mouse Homogentisate-1,2-Dioxygenase (HGD) ELISA Kit for detecting HGD in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O09173. Gene ID: 15233. Target Synonyms: AKU; HGO; Homogentisic Acid Oxidase; Homogentisicase; Homogentisate oxygenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3405