Learn More
Biomatik Corporation Human Lysine Specific Demethylase 3A (KDM3A) ELISA Kit
Supplier:  Biomatik Corporation EKU12062
Encompass_Preferred
Human Lysine Specific Demethylase 3A (KDM3A) ELISA Kit for detecting KDM3A in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y4C1. Gene ID: 55818. Target Synonyms: JMJD1; JMJD1A; TSGA; JHMD2A; JmjC domain-containing histone demethylation protein 2A; Jumonji domain-containing protein 1A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3411