Learn More
Biomatik Corporation Human Poly ADP Ribose Glycohydrolase (PARG) ELISA Kit
Supplier:  Biomatik Corporation EKU12099
Encompass_Preferred
Human Poly ADP Ribose Glycohydrolase (PARG) ELISA Kit for detecting PARG in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q86W56. Gene ID: 670. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3416