Learn More
Biomatik Corporation Human Leptin (LEP) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12118
Encompass_Preferred
Human Leptin (LEP) Mini sample ELISA Kit for detecting LEP in serum, plasma, saliva, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P41159. Gene ID: 3952. Target Synonyms: OB; OBS; Obesity Homolog; Obesity Factor; Obese Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3423