Learn More
Biomatik Corporation Rabbit Tumor Necrosis Factor Alpha (TNFa) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU12120
Encompass_Preferred
Rabbit Tumor Necrosis Factor Alpha (TNFa) Wide-range ELISA Kit for detecting TNFa in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04924. Gene ID: 100009088. Target Synonyms: DIF; TNF-A; TNFSF2; Cachectin; Tumor Necrosis Factor Ligand Superfamily Member 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3425