Learn More
Biomatik Corporation Mouse Heterogeneous Nuclear Ribonucleoprotein A2/B1 (HNRPA2B1) ELISA Kit
Supplier:  Biomatik Corporation EKU12125
Encompass_Preferred
Mouse Heterogeneous Nuclear Ribonucleoprotein A2/B1 (HNRPA2B1) ELISA Kit for detecting HNRPA2B1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88569. Gene ID: 53379. Target Synonyms: hnRNP-A2/B1; hnRNPA2B1; RA33; HnRNPA2/B1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3427