Learn More
Biomatik Corporation Pig Platelet/Endothelial Cell Adhesion Molecule (PECAM1) ELISA Kit
Supplier:  Biomatik Corporation EKU12128
Encompass_Preferred
Pig Platelet/Endothelial Cell Adhesion Molecule (PECAM1) ELISA Kit for detecting PECAM1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.053ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q95242. Gene ID: 396941. Target Synonyms: CD31; EndoCAM; GPIIA'; PECA1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3429