Learn More
Biomatik Corporation Mouse B-Lymphocyte Activation Antigen B7-1 (LAB7-1) ELISA Kit
Supplier:  Biomatik Corporation EKU12173
Encompass_Preferred
Mouse B-Lymphocyte Activation Antigen B7-1 (LAB7-1) ELISA Kit for detecting LAB7-1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q00609. Gene ID: 12519. Target Synonyms: CD80; CD28LG; CD28LG1; BB1; B7-1 Antigen; T-lymphocyte activation antigen CD80; CTLA-4 counter-receptor B7.1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3442