Learn More
Biomatik Corporation Human Latent Transforming Growth Factor Beta Binding Protein 1 (LTBP1) ELISA Kit
Supplier:  Biomatik Corporation EKU12193
Encompass_Preferred
Human Latent Transforming Growth Factor Beta Binding Protein 1 (LTBP1) ELISA Kit for detecting LTBP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q14766. Gene ID: 4052. Target Synonyms: TGF-beta1-BP-1; Transforming growth factor beta-1-binding protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3449