Learn More
Biomatik Corporation Human Grancalcin (GCA) ELISA Kit
Supplier:  Biomatik Corporation EKU12204
Encompass_Preferred
Human Grancalcin (GCA) ELISA Kit for detecting GCA in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P28676. Gene ID: 25801. Target Synonyms: GCL; EF Hand Calcium Binding Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3452