Learn More
Biomatik Corporation Rat Diazepam Binding Inhibitor (DBI) ELISA Kit
Supplier:  Biomatik Corporation EKU12217
Encompass_Preferred
Rat Diazepam Binding Inhibitor (DBI) ELISA Kit for detecting DBI in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500ng/ml. Sensitivity: 3.5ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11030. Gene ID: 25045. Target Synonyms: ACBP; ACBD1; CCK-RP; EP; Endozepine; GABA Receptor Modulator; Acyl-Coenzyme A Binding Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3454