Learn More
Biomatik Corporation Homocysteine (HCy) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12220
Encompass_Preferred
Homocysteine (HCy) Mini sample ELISA Kit for detecting HCy in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 98.77-8,000ng/ml. Sensitivity: 33.52ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3456