Learn More
Biomatik Corporation Human Activin A Receptor Type II B (ACVR2B) ELISA Kit
Supplier:  Biomatik Corporation EKU12301
Encompass_Preferred
Human Activin A Receptor Type II B (ACVR2B) ELISA Kit for detecting ACVR2B in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13705. Gene ID: 93. Target Synonyms: ACVR2-B; ActR-IIB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3485