Learn More
Biomatik Corporation Human Transient Receptor Potential Cation Channel Subfamily V, Member 6 (TRPV6) ELISA Kit
Supplier:  Biomatik Corporation EKU12332
Encompass_Preferred
Human Transient Receptor Potential Cation Channel Subfamily V, Member 6 (TRPV6) ELISA Kit for detecting TRPV6 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H1D0. Gene ID: 55503. Target Synonyms: CAT1; CATL; ABP/ZF; ECAC2; ZFAB; Epithelial Calcium Channel 2; CaT-like; Calcium transport protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3492