Learn More
Biomatik Corporation Human Protein Kinase, AMP Activated Beta 2 (PRKAb2) ELISA Kit
Supplier:  Biomatik Corporation EKU12341
Encompass_Preferred
Human Protein Kinase, AMP Activated Beta 2 (PRKAb2) ELISA Kit for detecting PRKAb2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 32pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O43741. Gene ID: 5565. Target Synonyms: 5'-AMP-activated protein kinase subunit beta-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3494