Learn More
Biomatik Corporation Rat Complement Component 3 (C3) CLIA Kit
Supplier:  Biomatik Corporation EKU12353
Encompass_Preferred
Rat Complement Component 3 (C3) CLIA Kit for detecting C3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.27-200ng/ml. Sensitivity: 0.12ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P01026. Gene ID: 24232. Target Synonyms: ASP; CPAMD1; Complement Protein C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a anaphylatoxin; Acylation stimulating protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3498