Learn More
Biomatik Corporation Human Asprosin (Asprosin) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12366
Encompass_Preferred
Human Asprosin (Asprosin) Mini sample ELISA Kit for detecting Asprosin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35555. Gene ID: 2200. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3500