Learn More
Biomatik Corporation Human Interleukin 17 (IL17) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU12392
Encompass_Preferred
Human Interleukin 17 (IL17) High-sensitive ELISA Kit for detecting IL17 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 3.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q16552. Gene ID: 3605. Target Synonyms: IL17A; CTLA8; IL-17A; Cytotoxic T-Lymphocyte-Associated Protein 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3511