Learn More
Biomatik Corporation Sheep Heat Shock Protein 90kDa Beta 1 (HSP90b1) ELISA Kit
Supplier:  Biomatik Corporation EKU12401
Encompass_Preferred
Sheep Heat Shock Protein 90kDa Beta 1 (HSP90b1) ELISA Kit for detecting HSP90b1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Sheep (Ovis aries). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: W5Q4E1. Target Synonyms: HSP90-B1; ECGP; GP96; GRP94; TRA1; Glycoprotein 96; Tumor Rejection Antigen (gp96) 1;94 kDa glucose-regulated protein; Endoplasmin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3516