Learn More
Biomatik Corporation Mouse Cholecystokinin (CCK) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12431
Encompass_Preferred
Mouse Cholecystokinin (CCK) Mini sample ELISA Kit for detecting CCK in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.09pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09240. Gene ID: 12424. Target Synonyms: CCK-PZ. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3521