Learn More
Biomatik Corporation Mouse Myosin Binding Protein C, Slow Type (MYBPC1) CLIA Kit
Supplier:  Biomatik Corporation EKU12479
Encompass_Preferred
Mouse Myosin Binding Protein C, Slow Type (MYBPC1) CLIA Kit for detecting MYBPC1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q6P6L5. Gene ID: 109272. Target Synonyms: MYBPCC; MYBPCS; C-protein, skeletal muscle slow isoform. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3531