Learn More
Biomatik Corporation Human 150 kDa Oxygen Regulated Protein (ORP150) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12512
Encompass_Preferred
Human 150 kDa Oxygen Regulated Protein (ORP150) Anti-antigen ELISA Kit for detecting ORP150 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.14ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: Q9Y4L1. Gene ID: 10525. Target Synonyms: HYOU1; HSP12A; Grp170; Glucose-Regulated Protein 170; Hypoxia Up Regulated 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3540