Learn More
Biomatik Corporation Dog Trefoil Factor 3 (TFF3) ELISA Kit
Supplier:  Biomatik Corporation EKU12529
Encompass_Preferred
Dog Trefoil Factor 3 (TFF3) ELISA Kit for detecting TFF3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q863B4. Gene ID: 403488. Target Synonyms: HITF; ITF3; TFI; HP1.B; Trefoil Factor 3, Intestinal. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3544