Learn More
Biomatik Corporation Bovine Vitamin A (Va) Elisa Kit
Supplier:  Biomatik Corporation EKC31023
Encompass_Preferred
Bovine Vitamin A (Va) Elisa Kit for detecting Vitamin A (Va) in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 6.25-100ng/ml. Sensitivity: 4.672ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3553