Learn More
Biomatik Corporation Cortisol (Cor) Elisa Kit
Supplier:  Biomatik Corporation EKC31045
Encompass_Preferred
Cortisol (Cor) Elisa Kit for detecting Cortisol (Cor) in serum, plasma, tissue homogenates. Species Reactivity: Universal. Detection Range: 0.049-200ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3555