Learn More
Biomatik Corporation Guinea Pig Albumin (Alb) Elisa Kit
Supplier:  Biomatik Corporation EKC31091
Encompass_Preferred
Guinea Pig Albumin (Alb) Elisa Kit for detecting Albumin in serum, plasma, tissue homogenates. Species Reactivity: Guinea pig (Cavia porcellus). Detection Range: 0.156-10ug/ml. Sensitivity: 0.09ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q6WDN9; Alb. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3562